siRNA Details
| Gene Symbol: Rac2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009008.3 | NP_033034.1 | 7 | 1145 | ||
| GenBank mRNA | AK007561.1 | - | 7 | 1083 | ||
| AK088415.1 | - | 7 | 1094 | |||
| AK088805.1 | - | 7 | 1082 | |||
| AK144136.1 | - | 7 | 1089 | |||
| AK171321.1 | - | 7 | 1146 | |||
| BC005455.1 | - | 7 | 1061 | 3495818 | ||
| X53247.1 | - | 7 | 1041 | |||

Rac2
Genomic Map