siRNA Details
| Gene Symbol: Rac2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009008.3 | NP_033034.1 | 7 | 1231 | ||
| GenBank mRNA | AK007561.1 | - | 7 | 1169 | ||
| AK011045.1 | - | 7 | 1213 | |||
| AK088415.1 | - | 7 | 1180 | |||
| AK088805.1 | - | 7 | 1168 | |||
| AK144136.1 | - | 7 | 1175 | |||
| AK171321.1 | - | 7 | 1232 | |||
| BC005455.1 | - | 7 | 1147 | 3495818 | ||
| X53247.1 | - | 7 | 1127 | |||

Rac2
Genomic Map