siRNA Details
| Gene Symbol: Rad17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001044371.2 | NP_001037836.1 | 15 | 1882 | ||
| NM_001283011.1 | NP_001269940.1 | 16 | 2087 | |||
| NM_011233.3 | NP_035363.2 | 15 | 2010 | |||
| GenBank mRNA | AF085737.1 | - | 15 | 1933 | ||
| AJ011923.1 | - | 14 | 1783 | |||
| AK034773.1 | - | 1 | 1785 | |||
| AK134277.1 | - | 15 | 1837 | |||
| AK162016.1 | - | 15 | 1870 | |||
| AK167358.1 | - | 16 | 2030 | |||
| BC050267.1 | - | 12 | 1522 | |||
| BC066855.1 | - | 15 | 2000 | 30473984 | ||

Rad17
Genomic Map