siRNA Details
| Gene Symbol: Rad17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001044371.2 | NP_001037836.1 | 10 | 1145 | ||
| NM_001283011.1 | NP_001269940.1 | 11 | 1350 | |||
| NM_011233.3 | NP_035363.2 | 10 | 1273 | |||
| GenBank mRNA | AF085737.1 | - | 10 | 1196 | ||
| AJ011923.1 | - | 9 | 1046 | |||
| AK134277.1 | - | 10 | 1100 | |||
| AK162016.1 | - | 10 | 1133 | |||
| AK167358.1 | - | 11 | 1293 | |||
| AK209359.1 | - | 2 | 92 | |||
| BC023939.1 | - | 10 | 1150 | |||
| BC050267.1 | - | 7 | 785 | |||
| BC066855.1 | - | 10 | 1263 | 30473984 | ||

Rad17
Genomic Map