siRNA Details
| Gene Symbol: Rad50 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009012.2 | NP_033038.2 | 10 | 1756 | ||
| GenBank mRNA | AK037641.1 | - | 10 | 1757 | ||
| AK087982.1 | - | 2 | 167 | |||
| U66887.1 | - | 10 | 1734 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rad50| Gene Symbol: Rad50 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009012.2 | NP_033038.2 | 10 | 1756 | ||
| GenBank mRNA | AK037641.1 | - | 10 | 1757 | ||
| AK087982.1 | - | 2 | 167 | |||
| U66887.1 | - | 10 | 1734 | |||