siRNA Details
| Gene Symbol: Rad50 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009012.2 | NP_033038.2 | 20 | 3327 | ||
| GenBank mRNA | U66887.1 | - | 20 | 3305 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rad50| Gene Symbol: Rad50 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009012.2 | NP_033038.2 | 20 | 3327 | ||
| GenBank mRNA | U66887.1 | - | 20 | 3305 | ||