siRNA Details
| Gene Symbol: Rad51ap1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009013.3 | NP_033039.2 | 3 | 212 | ||
| GenBank mRNA | AK011379.1 | - | 3 | 213 | ||
| AK077829.1 | - | 3 | 245 | |||
| AK079525.1 | - | 3 | 234 | |||
| AK166703.1 | - | 3 | 245 | |||
| BC003738.1 | - | 3 | 226 | 3495632 | ||
| U93583.1 | - | 3 | 210 | |||

Rad51ap1
Genomic Map