siRNA Details
| Gene Symbol: Rad51b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001252562.1 | NP_001239491.1 | 3, 4 | 261 | ||
| NM_009014.3 | NP_033040.2 | 3, 4 | 279 | |||
| GenBank mRNA | AK015771.1 | - | 3, 4 | 261 | ||
| AK032913.1 | - | 3, 4 | 280 | |||
| U92068.1 | - | 2, 3 | 221 | |||

Rad51b
Genomic Map