siRNA Details
| Gene Symbol: Rad9a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011237.2 | NP_035367.1 | 10 | 999 | ||
| GenBank mRNA | AF045663.1 | - | 10 | 999 | ||
| AK134293.1 | - | 10 | 1012 | |||
| AK138973.1 | - | 9 | 1077 | |||
| AK165833.1 | - | 10 | 1035 | |||
| AK179097.1 | - | 4 | 386 | |||
| BC082556.1 | - | 10 | 938 | 30538342 | ||

Rad9a
Genomic Map