siRNA Details
| Gene Symbol: Rag1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009019.2 | NP_033045.2 | 2 | 465 | ||
| GenBank mRNA | AK088725.1 | - | 2 | 466 | ||
| AK162974.1 | - | 2 | 465 | |||
| BC138342.1 | - | 2 | 390 | 8861362 | ||
| M29475.1 | - | 2 | 434 | |||

Rag1
Genomic Map