siRNA Details
| Gene Symbol: Rag2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009020.3 | NP_033046.1 | 3 | 467 | ||
| GenBank mRNA | AK031014.1 | - | 3 | 477 | ||
| AK040375.1 | - | 3 | 468 | |||
| AK089075.1 | - | 3 | 499 | |||
| BC120548.1 | - | 3 | 424 | 40129471 | ||
| BC125473.1 | - | 3 | 424 | 40129988 | ||
| BC144856.1 | - | 2 | 361 | 9053404 | ||
| M64796.1 | - | 3 | 411 | |||

Rag2
Genomic Map