siRNA Details
| Gene Symbol: Rasgrp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011246.2 | NP_035376.1 | 9 | 1303 | ||
| GenBank mRNA | AF106070.1 | - | 9 | 1147 | ||
| AK028308.1 | - | 2 | 294 | |||
| AK047613.1 | - | 4 | 605 | |||
| AK134005.1 | - | 9 | 1317 | |||
| AK141524.1 | - | 9 | 1428 | |||
| BC057120.1 | - | 9 | 1303 | 6831137 | ||
| BC057341.1 | - | 9 | 1303 | 6831137 | ||

Rasgrp1
Genomic Map