siRNA Details
| Gene Symbol: Rbm6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011251.3 | NP_035381.2 | 9, 10 | 2207 | ||
| NM_029169.3 | NP_083445.1 | 9, 10 | 2478 | |||
| GenBank mRNA | AJ006486.2 | - | 9, 10 | 2086 | ||
| AK032150.1 | - | 4, 5 | 459 | |||
| AK077899.1 | - | 9, 10 | 2385 | |||
| AK145612.1 | - | 9, 10 | 2367 | |||
| AK164432.1 | - | 4, 5 | 564 | |||
| AK220446.1 | - | 9, 10 | 2356 | |||
| BC026129.1 | - | 9, 10 | 2232 | 3982861 | ||
| BC026130.1 | - | 9, 10 | 2381 | |||

Rbm6
Genomic Map