siRNA Details
| Gene Symbol: Rbpms | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001042674.2 | NP_001036139.1 | 2, 3 | 475 | ||
| NM_001042675.2 | NP_001036140.1 | 2, 3 | 475 | |||
| NM_001286168.1 | NP_001273097.1 | 2, 3 | 475 | |||
| NM_019733.3 | NP_062707.1 | 2, 3 | 475 | |||
| GenBank mRNA | AF148511.1 | - | 2, 3 | 424 | ||
| AK012265.1 | - | 2, 3 | 418 | |||
| AK012586.1 | - | 2, 3 | 418 | |||
| AK041118.1 | - | 2, 3 | 419 | |||
| AK079238.1 | - | 2, 3 | 418 | |||
| BB637557.1 | - | 2, 3 | 419 | |||
| BC030397.1 | - | 2, 3 | 436 | 4225078 | ||
| CK030989.1 | - | 2, 3 | 453 | |||

Rbpms
Genomic Map