siRNA Details
| Gene Symbol: Rbpjl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009036.1 | NP_033062.1 | 3 | 291 | ||
| GenBank mRNA | AK052449.1 | - | 3 | 1417 | ||
| AK131814.1 | - | 3 | 272 | |||
| AK131832.1 | - | 3 | 277 | |||
| BC152311.1 | - | 3 | 285 | 40090714 | ||
| Y10926.1 | - | 3 | 291 | |||

Rbpjl
Genomic Map