siRNA Details
| Gene Symbol: Rfc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011258.2 | NP_035388.2 | 22 | 2980 | ||
| GenBank mRNA | AK134660.1 | - | 7 | 758 | ||
| AK143077.1 | - | 3 | 1512 | |||
| BC085173.1 | - | 22 | 2957 | 30549691 | ||
| BC085174.1 | - | 22 | 2947 | |||
| U01222.1 | - | 22 | 2948 | |||
| U07157.1 | - | 22 | 2943 | |||
| U36441.1 | - | 22 | 2954 | |||
| X72711.1 | - | 22 | 2946 | |||

Rfc1
Genomic Map