siRNA Details
| Gene Symbol: Reg2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009043.1 | NP_033069.1 | 6 | 550 | ||
| GenBank mRNA | AK008196.1 | - | 6 | 549 | ||
| BC098503.1 | - | 5 | 533 | 6431377 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Reg2| Gene Symbol: Reg2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009043.1 | NP_033069.1 | 6 | 550 | ||
| GenBank mRNA | AK008196.1 | - | 6 | 549 | ||
| BC098503.1 | - | 5 | 533 | 6431377 | ||