siRNA Details
| Gene Symbol: Rela | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009045.4 | NP_033071.1 | 11 | 1609 | ||
| GenBank mRNA | AK089489.1 | - | 11 | 1354 | ||
| AK144565.1 | - | 11 | 1356 | |||
| AK154630.1 | - | 11 | 1354 | |||
| AK155321.1 | - | 11 | 1618 | |||
| AK155889.1 | - | 11 | 1351 | |||
| AK157076.1 | - | 11 | 1341 | |||
| AK158072.1 | - | 11 | 1619 | |||
| AK212853.1 | - | 1 | 27 | |||
| BC003818.1 | - | 11 | 1368 | 3489295 | ||
| BC094053.1 | - | 11 | 1609 | 3157927 | ||
| DQ020177.1 | - | 9 | 1178 | |||
| M61909.1 | - | 11 | 1324 | |||

Rela
Genomic Map