siRNA Details
| Gene Symbol: Reln | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011261.2 | NP_035391.2 | 20 | 2909 | ||
| GenBank mRNA | AK082447.1 | - | 20 | 2910 | ||
| AY704216.1 | - | 1 | 20 | |||
| U24703.1 | - | 20 | 2894 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Reln| Gene Symbol: Reln | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011261.2 | NP_035391.2 | 20 | 2909 | ||
| GenBank mRNA | AK082447.1 | - | 20 | 2910 | ||
| AY704216.1 | - | 1 | 20 | |||
| U24703.1 | - | 20 | 2894 | |||