siRNA Details
| Gene Symbol: Upf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001122829.1 | NP_001116301.1 | 23 | 3549 | ||
| NM_030680.2 | NP_109605.2 | 23 | 3516 | |||
| GenBank mRNA | AF182947.1 | - | 24 | 3505 | ||
| AF322655.1 | - | 23 | 3236 | |||
| AK052517.1 | - | 12 | 1584 | |||
| AK148196.1 | - | 24 | 3526 | |||
| BC030916.1 | - | 11 | 1527 | |||
| BC052149.1 | - | 23 | 3454 | 6332288 | ||
| BC056442.1 | - | 23 | 3549 | 5702493 | ||

Upf1
Genomic Map