siRNA Details
| Gene Symbol: Resp18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009049.1 | NP_033075.1 | 5 | 506 | ||
| GenBank mRNA | AK019440.1 | - | 5 | 456 | ||
| BC027535.1 | - | 5 | 469 | 3376899 | ||
| FJ621497.1 | - | 4 | 399 | |||
| L34214.1 | - | 5 | 506 | |||

Resp18
Genomic Map