siRNA Details
| Gene Symbol: Resp18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009049.1 | NP_033075.1 | 3 | 310 | ||
| GenBank mRNA | AK012477.1 | - | 3 | 263 | ||
| AK019440.1 | - | 3 | 260 | |||
| BC027535.1 | - | 3 | 273 | 3376899 | ||
| FJ621497.1 | - | 3 | 246 | |||
| L34214.1 | - | 3 | 310 | |||

Resp18
Genomic Map