siRNA Details
| Gene Symbol: Rest | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011263.2 | NP_035393.2 | 3 | 1204 | ||
| GenBank mRNA | AB024496.1 | - | 2 | 900 | ||
| AB221589.1 | - | 2 | 899 | |||
| AK004945.2 | - | 3 | 1207 | |||
| AK156514.1 | - | 3 | 1190 | |||
| BC096434.1 | - | 3 | 1127 | |||
| BC127065.1 | - | 2 | 900 | 40110015 | ||
| DQ644039.1 | - | 2 | 900 | |||
| JN019832.1 | - | 2 | 900 | |||
| U13878.1 | - | 2 | 675 | |||

Rest
Genomic Map