siRNA Details
| Gene Symbol: Bex1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009052.2 | NP_033078.2 | 3 | 447 | ||
| GenBank mRNA | AF051347.1 | - | 3 | 380 | ||
| AF097438.1 | - | 3 | 392 | |||
| AK003429.1 | - | 1 | 62 | |||
| BC055751.1 | - | 210 | ||||
| BC058805.1 | - | 1 | 719 | |||
| BC061179.1 | - | 3 | 392 | 6531808 | ||
| BC089464.1 | - | 3 | 441 | 6594252 | ||
| CB194115.1 | - | 3 | 369 | |||

Bex1
Genomic Map