siRNA Details
| Gene Symbol: Rfx3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001166414.1 | NP_001159886.1 | 5 | 568 | ||
| NM_011265.3 | NP_035395.2 | 4 | 609 | |||
| GenBank mRNA | AK030008.1 | - | 6 | 641 | ||
| AK041120.1 | - | 5 | 669 | |||
| AK140928.1 | - | 4 | 492 | |||
| BC017598.1 | - | 4 | 583 | 4483833 | ||

Rfx3
Genomic Map