siRNA Details
| Gene Symbol: Slc50a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009057.3 | NP_033083.2 | 3 | 313 | ||
| GenBank mRNA | AK014305.1 | - | 3 | 266 | ||
| AK045924.1 | - | 1 | 626 | |||
| AK079716.1 | - | 2 | 378 | |||
| AK158862.1 | - | 2 | 338 | |||
| AK170162.1 | - | 3 | 264 | |||
| AK177059.1 | - | 3 | 221 | |||
| BC014292.1 | - | 3 | 297 | 4190124 | ||
| BY333933.1 | - | 3 | 184 | |||
| X96618.1 | - | 3 | 304 | |||

Slc50a1
Genomic Map