siRNA Details
| Gene Symbol: IL3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_000588.3 | NP_000579.2 | 5 | 434 | ||
| GenBank mRNA | BC066272.1 | - | 5 | 427 | 6971773 | |
| BC066273.1 | - | 5 | 427 | 6971774 | ||
| BC066274.1 | - | 5 | 427 | 6971775 | ||
| BC066275.1 | - | 5 | 427 | 6971776 | ||
| BC066276.1 | - | 5 | 427 | 6971777 | ||
| BC069472.1 | - | 5 | 430 | 7262122 | ||
| M14743.1 | - | 5 | 390 | |||
| M17115.1 | - | 5 | 419 | |||
| M20137.1 | - | 5 | 434 | |||

IL3
Genomic Map