siRNA Details
| Gene Symbol: Rgs16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011267.3 | NP_035397.2 | 2 | 203 | ||
| GenBank mRNA | AK079543.1 | - | 2 | 204 | ||
| AK154324.1 | - | 2 | 202 | |||
| AK193821.1 | - | 1 | 24 | |||
| BC010980.1 | - | 2 | 196 | 4020813 | ||
| BC049968.1 | - | 2 | 191 | 4021479 | ||
| BC055459.1 | - | 2 | 207 | 4194934 | ||
| U67189.1 | - | 2 | 193 | |||
| U72881.1 | - | 2 | 158 | |||
| U94828.1 | - | 2 | 196 | |||

Rgs16
Genomic Map