siRNA Details
| Gene Symbol: IL3RA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_002183.3 | NP_002174.1 | 3 | 430 | ||
| XM_005274431.4 | XP_005274488.1 | 3 | 467 | |||
| XM_005274432.1 | XP_005274489.1 | 3 | 260 | |||
| XM_017029491.1 | XP_016884980.1 | 3 | 468 | |||
| GenBank mRNA | AK290568.1 | - | 3 | 260 | ||
| BC035407.1 | - | 3 | 430 | 5167281 | ||
| M74782.1 | - | 3 | 227 | |||

IL3RA
Genomic Map