siRNA Details
| Gene Symbol: Ripk1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009068.3 | NP_033094.3 | 5 | 706 | ||
| GenBank mRNA | AK030959.1 | - | 5 | 937 | ||
| AK156803.1 | - | 5 | 971 | |||
| BC050905.1 | - | 5 | 706 | 5721177 | ||
| BC054542.1 | - | 5 | 706 | 5721177 | ||
| BC058162.1 | - | 5 | 892 | 5135235 | ||
| U25995.1 | - | 4 | 621 | |||

Ripk1
Genomic Map