siRNA Details
| Gene Symbol: Rlim | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011276.3 | NP_035406.3 | 5 | 1124 | ||
| GenBank mRNA | AF069992.1 | - | 5 | 1079 | ||
| AK013207.1 | - | 5 | 1096 | |||
| AK029295.1 | - | 5 | 1118 | |||
| BC012960.1 | - | 3 | 857 | 4193003 | ||

Rlim
Genomic Map