siRNA Details
| Gene Symbol: Trim10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011280.2 | NP_035410.2 | 3, 4 | 1069 | ||
| GenBank mRNA | AF134811.1 | - | 3, 4 | 1051 | ||
| AF220121.1 | - | 3, 4 | 1056 | |||
| AK011082.1 | - | 3, 4 | 1069 | |||
| BC051632.1 | - | 3, 4 | 1042 | 6507331 | ||

Trim10
Genomic Map