siRNA Details
| Gene Symbol: Rnps1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001080127.1 | NP_001073596.1 | 5, 6 | 697 | ||
| NM_001080128.1 | NP_001073597.1 | 4, 5 | 516 | |||
| NM_009070.2 | NP_033096.2 | 6, 7 | 819 | |||
| GenBank mRNA | AK011475.1 | - | 5, 6 | 699 | ||
| AK146443.1 | - | 4, 5 | 515 | |||
| AK151150.1 | - | 5, 6 | 687 | |||
| AK165905.1 | - | 4, 5 | 503 | |||
| BC002061.1 | - | 5, 6 | 685 | 3485311 | ||
| BC008089.1 | - | 2, 3 | 254 | |||
| X70067.1 | - | 5, 6 | 704 | |||

Rnps1
Genomic Map