siRNA Details
| Gene Symbol: Rock1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009071.2 | NP_033097.1 | 19 | 2745 | ||
| GenBank mRNA | AK085974.1 | - | 6 | 859 | ||
| BC066873.1 | - | 19 | 2422 | |||
| U58512.1 | - | 19 | 2270 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rock1| Gene Symbol: Rock1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009071.2 | NP_033097.1 | 19 | 2745 | ||
| GenBank mRNA | AK085974.1 | - | 6 | 859 | ||
| BC066873.1 | - | 19 | 2422 | |||
| U58512.1 | - | 19 | 2270 | |||