siRNA Details
| Gene Symbol: Rom1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009073.4 | NP_033099.3 | 2 | 791 | ||
| GenBank mRNA | AK133529.1 | - | 2 | 792 | ||
| BC014827.1 | - | 2 | 613 | 4527997 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rom1| Gene Symbol: Rom1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009073.4 | NP_033099.3 | 2 | 791 | ||
| GenBank mRNA | AK133529.1 | - | 2 | 792 | ||
| BC014827.1 | - | 2 | 613 | 4527997 | ||