siRNA Details
| Gene Symbol: Ros1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011282.2 | NP_035412.2 | 10 | 1276 | ||
| GenBank mRNA | X81650.1 | - | 10 | 1099 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ros1| Gene Symbol: Ros1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011282.2 | NP_035412.2 | 10 | 1276 | ||
| GenBank mRNA | X81650.1 | - | 10 | 1099 | ||