siRNA Details
| Gene Symbol: Rp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011283.2 | NP_035413.1 | 4 | 5783 | ||
| GenBank mRNA | AF146593.2 | - | 1 | 2449 | ||
| AF155141.2 | - | 4 | 5774 | |||
| BC028267.1 | - | 1 | 2333 | |||
| BC038794.1 | - | 4 | 5725 | |||
| BC061512.1 | - | 1 | 3004 | |||
| BC096607.1 | - | 4 | 5783 | |||
| BC120927.1 | - | 4 | 5737 | 40099705 | ||
| BC120928.1 | - | 4 | 5737 | 40099706 | ||

Rp1
Genomic Map