siRNA Details
| Gene Symbol: Rpe65 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029987.2 | NP_084263.2 | 14 | 1558 | ||
| GenBank mRNA | AF410461.1 | - | 13 | 1506 | ||
| BC090656.1 | - | 14 | 1534 | |||
| BC130028.1 | - | 13 | 1510 | 40140883 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rpe65| Gene Symbol: Rpe65 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029987.2 | NP_084263.2 | 14 | 1558 | ||
| GenBank mRNA | AF410461.1 | - | 13 | 1506 | ||
| BC090656.1 | - | 14 | 1534 | |||
| BC130028.1 | - | 13 | 1510 | 40140883 | ||