siRNA Details
| Gene Symbol: Rph3a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AB112924.1 | - | 21, 22 | 2155 | ||
| AK139040.1 | - | 20, 21 | 2099 | |||
| BC042585.1 | - | 21, 22 | 2322 | 5364086 | ||
| BC050883.1 | - | 21, 22 | 2289 | 5698459 | ||
| D29965.1 | - | 21, 22 | 2132 | |||

Rph3a
Genomic Map