siRNA Details
| Gene Symbol: Rpl10a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011287.2 | NP_035417.2 | 5 | 599 | ||
| GenBank mRNA | AK150743.1 | - | 5 | 487 | ||
| AK151172.1 | - | 5 | 488 | |||
| AK153863.1 | - | 5 | 485 | |||
| BC006039.1 | - | 3 | 816 | 3494193 | ||
| BC083346.1 | - | 4 | 465 | 5712034 | ||
| D10916.1 | - | 2 | 182 | |||
| U12403.1 | - | 5 | 471 | |||

Rpl10a
Genomic Map