siRNA Details
Gene Symbol: Rpl28 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009081.2 | NP_033107.1 | 2 | 106 | ||
GenBank mRNA | AK012368.1 | - | 2 | 107 | ||
AK018714.1 | - | 2 | 107 | |||
AK076133.1 | - | 2 | 107 | |||
BC024395.1 | - | 2 | 106 | 4981342 | ||
BC086797.1 | - | 2 | 92 | 5713507 | ||
BC086932.1 | - | 2 | 108 | 6760499 | ||
BC092012.1 | - | 2 | 106 | 30608657 | ||
X74856.1 | - | 2 | 134 |

More Information
Other Species Targeted

Panther Classification:
Gene Ontology Categories:
