siRNA Details
| Gene Symbol: Polr1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009086.2 | NP_033112.2 | 15 | 3007 | ||
| GenBank mRNA | AK083732.1 | - | 15 | 2990 | ||
| AK143501.1 | - | 15 | 2944 | |||
| AK144616.1 | - | 15 | 2994 | |||
| AK156712.1 | - | 15 | 2997 | |||
| BC060656.1 | - | 15 | 2937 | 30355130 | ||
| U58280.1 | - | 15 | 2971 | |||

Polr1b
Genomic Map