siRNA Details
| Gene Symbol: Rps17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009092.3 | NP_033118.1 | 3 | 241 | ||
| GenBank mRNA | AK007992.1 | - | 3 | 249 | ||
| AK010647.1 | - | 3 | 243 | |||
| AK012781.1 | - | 3 | 230 | |||
| AK158902.1 | - | 2 | 307 | |||
| AK162022.1 | - | 3 | 235 | |||
| AK162335.1 | - | 3 | 233 | |||
| AK167206.1 | - | 3 | 235 | |||
| BC081466.1 | - | 2 | 228 | 6412006 | ||
| BC086900.1 | - | 3 | 234 | 6760363 | ||
| BC086901.1 | - | 3 | 234 | 6763239 | ||
| D25213.1 | - | 2 | 222 | |||

Rps17
Genomic Map