siRNA Details
| Gene Symbol: Rras | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009101.2 | NP_033127.1 | 5 | 506 | ||
| GenBank mRNA | AK155855.1 | - | 5 | 536 | ||
| BC009105.1 | - | 5 | 523 | 3483313 | ||
| BC110562.1 | - | 5 | 508 | 40049793 | ||
| BC110563.1 | - | 5 | 508 | 40049794 | ||
| BC145932.1 | - | 5 | 552 | 40131235 | ||
| BC145934.1 | - | 5 | 552 | 40131237 | ||
| M21019.1 | - | 5 | 508 | |||

Rras
Genomic Map