siRNA Details
| Gene Symbol: Rrh | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009102.3 | NP_033128.1 | 3 | 375 | ||
| GenBank mRNA | AF012271.1 | - | 3 | 379 | ||
| AK044717.1 | - | 363 | ||||
| BC046288.1 | - | 4 | 501 | 6468540 | ||
More Information
Set Membership: |
|

Rrh
Genomic Map