siRNA Details
| Gene Symbol: Rrh | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009102.3 | NP_033128.1 | 4 | 435 | ||
| GenBank mRNA | AF012271.1 | - | 4 | 439 | ||
| AK021379.1 | - | 2 | 145 | |||
| AK044717.1 | - | 423 | ||||
| BC046288.1 | - | 5 | 561 | 6468540 | ||
More Information
Other Species Targeted
Set Membership: |
|

Rrh
Genomic Map