siRNA Details
| Gene Symbol: Rsu1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009105.4 | NP_033131.2 | 8 | 920 | ||
| GenBank mRNA | AK009288.1 | - | 8 | 810 | ||
| AK011857.1 | - | 8 | 813 | |||
| AK013076.1 | - | 4 | 391 | |||
| AK014749.1 | - | 3 | 263 | |||
| AK135491.1 | - | 9 | 903 | |||
| AK149835.1 | - | 8 | 791 | |||
| AK150011.1 | - | 8 | 788 | |||
| AK152655.1 | - | 8 | 800 | |||
| X63039.1 | - | 8 | 735 | |||

Rsu1
Genomic Map