siRNA Details
| Gene Symbol: Rtn2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025364.3 | NP_001020535.1 | 4, 5 | 528 | ||
| NM_013648.6 | NP_038676.1 | 7, 8 | 1442 | |||
| GenBank mRNA | AF038538.1 | - | 7, 8 | 1398 | ||
| AF038539.1 | - | 4, 5 | 467 | |||
| AK144271.1 | - | 7, 8 | 1405 | |||
| AK160500.1 | - | 4, 5 | 489 | |||
| BC031370.1 | - | 7, 8 | 1399 | 5359358 | ||

Rtn2
Genomic Map