siRNA Details
| Gene Symbol: Uimc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001293660.1 | NP_001280589.1 | 7, 8 | 741 | ||
| NM_011307.2 | NP_035437.1 | 9, 10 | 1579 | |||
| GenBank mRNA | AK052704.1 | - | 9, 10 | 1580 | ||
| AK083342.1 | - | 9, 10 | 1520 | |||
| AK160926.1 | - | 7, 8 | 742 | |||
| BC037092.1 | - | 8, 9 | 1454 | |||
| BC040808.1 | - | 9, 10 | 1535 | 4038778 | ||
| BC051659.1 | - | 9, 10 | 1502 | |||
| BC066220.1 | - | 9, 10 | 1502 | |||
| U22015.1 | - | 5, 6 | 1031 | |||

Uimc1
Genomic Map