siRNA Details
| Gene Symbol: Nr1h4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001163504.1 | NP_001156976.1 | 6 | 1074 | ||
| NM_001163700.1 | NP_001157172.1 | 5 | 870 | |||
| NM_009108.2 | NP_033134.2 | 5 | 858 | |||
| GenBank mRNA | AK002513.1 | - | 5 | 856 | ||
| AK132255.1 | - | 5 | 850 | |||
| BC015261.1 | - | 5 | 829 | 4241705 | ||
| U09416.1 | - | 5 | 827 | |||
| U09417.1 | - | 6 | 1038 | |||
| U09418.1 | - | 6 | 1035 | |||

Nr1h4
Genomic Map